Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EXOC7 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 70 kDa band observed in human lung lysates (calculated MW of 74.7 kDa according to NP_056034.2). Recommended concentration: 0.5-1.5 μg/mL. Immunohistochemistry: In paraffin embedded human adrenal gland shows staining of cytoplasm membranes and cytoplasm of secretory cells in the medulla. Recommended concentration, 5-10 μg/mL.

EXOC7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EXOC7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: EXOC7
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat
Reactivity: Human

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-557
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol EXOC7
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms EXOC1, EX070, Exo70p, YJL085W, EXO70, 2-5-3p
Concentration 500 ug/mL
Immunogen EXOC7 antibody was raised against a 12 amino acid synthetic peptide near the internal region of EXOC7.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo