Tlačiť

Anti-XPO7 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-558EA 614.75 EUR
PRSI45-558
Anti-XPO7 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1/32,000. Western Blot: Approximately 110 kDa band observed in human testis and Hela lysates (predicted size of approximately 124 kDa according to NP_055839). Recommended for use at 0.5-1 μg/mL

Exportin 7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Exportin 7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: XPO7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR