Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EYA4 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 50 kDa band in human and mouse brain lysates and in pituitary mouse cell line NIH 3T3 after 0.1 μg/mL antibody staining.5 kDa, 67.1 kDa and 69.4 kDa according to NP_004091.2, NP_742102.1 and NP_742103.1 respectively. The 50 kDa band was successfully blocked by incubation with the immunizing peptide.

EYA4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EYA4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EYA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse




 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-559
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol EYA4
Antigen name Eyes Absent homolog 4
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
ELISA Yes
Antigen synonyms DFNA10, CMD1J
Concentration 500 ug/mL
Immunogen EYA4 antibody was raised against a 15 amino acid synthetic peptide near the N-Terminus of EYA4.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo
 

Nový vzhľad a obnovený záväzok

Postavené na funkciách a zdrojoch, ktoré poznáte, optimalizované s inováciami od spoločnosti Avantor®. Naša nová značka symbolizuje agilitu a našu schopnosť prispôsobiť sa potrebám zákazníkov.

ĎALŠIE INFORMÁCIE ZAČAŤ NAKUPOVAŤ