Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-ACSL4 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 80 kDa band observed in HEPG2 lysate (calculated MW of 79.2 kDa according to NP_075266.1). Recommended concentration: 0.1-0.3 μg/mL.

FACL4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FACL4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: ACSL4
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat
Reactivity: Human

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-563
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol ACSL4
Antigen name Acyl-CoA Synthetase Long-chain Family Member 4
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
Antigen synonyms LACS4, ACS4, MRX68, MRX63, FACL4
Concentration 500 ug/mL
Immunogen FACL4 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of FACL4.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo