Tlačiť

Anti-FANCG Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-567EA 614.75 EUR
PRSI45-567
Anti-FANCG Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 70 kDa band observed in HeLa lysate (calculated MW of 68.6 kDa according to NP_004620.1). In transfected HEK293 transiently expressing FANCG a band of approximately 65 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.5-1 μg/mL.

FANCG antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FANCG antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FANCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR