Tlačiť

Anti-PLEKHA8 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-568EA 614.75 EUR
PRSI45-568
Anti-PLEKHA8 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:64,000. Immunohistochemistry: In paraffin embedded human placenta shows strong staining of cytoplasm in the margins of villi. Recommended concentration, 2-4 μg/mL.

FAPP2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FAPP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLEKHA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR