Tlačiť

Anti-KDM2B Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-574EA 616.08 EUR
PRSI45-574
Anti-KDM2B Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Customer's experiments gave bands at approximately 170+90 kDa in lysates of celll line HeLa (calculated MW of 153 kDa according to NP_115979.3). Recommended concentration: 1-3 μg/mL.

FBXL10 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FBXL10 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KDM2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR