Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-FBXL6 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:8,000.

FBXL6 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FBXL6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: FBXL6
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-579
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol FBXL6
Antigen name F-Box and Leucine-rich Repeat Protein 6
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Concentration 500 ug/mL
Immunogen FBXL6 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of FBXL6.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo