Tlačiť

Anti-FBXO44 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-581EA 614.75 EUR
PRSI45-581
Anti-FBXO44 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 60-65 kDa band in HepG2 and 293 lysates at 1 μg/mL. Further, this band was successfully blocked by incubation with the immunizing peptide.

FBXO44 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FBXO44 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FBXO44
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR