Tlačiť

Anti-FEM1C Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-585EA 616.08 EUR
PRSI45-585
Anti-FEM1C Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 75 kDa band observed in heart and kidney lysates from mouse and rat (calculated MW of 68.6 kDa according to mouse NP_775599.1). Recommended concentration: 0.3-1 μg/mL.

FEM1C antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FEM1C antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FEM1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR