Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-FGF23 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26-27 kDa band observed in human hippocampus lysates (calculated MW of 27.9 kDa according to NP_065689.1). Recommended concentration: 0.5-1.5 μg/mL. an additional band of unknown identity was also consistently observed at 37 kDa . This band was successfully blocked by incubation with the immunizing peptide.

FGF23 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FGF23 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FGF23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-588
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol FGF23
Antigen name Fibroblast Growth Factor 23
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ELISA Yes
Concentration 500 ug/mL
Immunogen FGF23 antibody was raised against a 13 amino acid synthetic peptide near the internal region of FGF23.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo