Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-FGFR1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 75 kDa band observed in human breast lysates (calculated MW of 82.2 kDa according to NP_075593, 81.9 kDa according to NP_075594). Recommended concentration: 0.3-1 μg/mL. Immunohistochemistry: In paraffin embedded human small intestine shows staining of the enterocytic cytoplasm. Recommended concentration, 4-6 μg/mL.

FGFR1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FGFR1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FGFR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-589
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol FGFR1
Antigen name Fibroblast Growth Factor Receptor 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms FLG, HH2, BFGFR, FGFBR, FLT-2, N-SAM, FGFR-1, CEK, HRTFDS, OGD, CD331, bFGF-R-1, FLT2, HBGFR, KAL2
Concentration 500 ug/mL
Immunogen FGFR1 antibody was raised against a 16 amino acid synthetic peptide near the C-Terminus of FGFR1.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo