Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-FOXA1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 50 kDa band observed in human HEPG2 lysate (calculated MW of 49.1 kDa according to NP_004487.2). Recommended concentration: 0.2-0.6 μg/mL. an additional band of unknown identity was also consistently observed at 35 kDa . This band was successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: Immunohistochemistry: In paraffin embedded human prostate shows nuclear staining of secretory cells. Recommended concentration, 5-10 μg/mL.

FOXA1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FOXA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: FOXA1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat
Reactivity: Human, Mouse

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-599
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol FOXA1
Antigen name Forkhead box protein A1
Reactivity Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ChIP Yes
Antigen synonyms Hnf3a, fkd7, tcf3a, Ci-fkh, FoxA4a, fkh, Hnf-3a, tFoxA1, HNF3A, xfkh2, Tcf3a, xfkh1, MGC52590, HNF3-alpha, HNF3alpha, foxa1, MGC75967, forkhead-7, Tcf-3a, MGC33105, pintallavis, FOXA1, HNF-3a, TCF3A
Concentration 500 ug/mL
Immunogen FOXA1 antibody was raised against an 11 amino acid synthetic peptide near the C-Terminus of FOXA1.
Form Liquid
Tested applications Chromatin
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo