Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-FOXC1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32,000. Approximately 55 kDa band observed in HEK293 lysate (calculated MW of 56.8 kDa according to NP_001444.2). Recommended concentration: 2-4 μg/mL.

FOXC1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FOXC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: FOXC1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat
Reactivity: Human

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-604
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol FOXC1
Antigen name Forkhead Box C1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Antigen synonyms IGDA, RIEG3, FREAC-3, ARA, FKHL7, IHG1, FREAC3, IRID1
Concentration 500 ug/mL
Immunogen FOXC1 antibody was raised against a 13 amino acid synthetic peptide near the internal region of FOXC1.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo