Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-FOXP2 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 85 kDa band observed in human brain (cerebellum and frontal cortex) lysates and in rat brain lysates (calculated MW of 82.6 kDa according to NP_683696.1). Recommended concentration: 0.3-1 μg/mL. an additional band of unknown identity was also consistently observed at 28-30 kDa . This band was successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: In paraffin embedded human prostate shows nuclear staining in the secretory cells. Recommended concentration, 2-4 μg/mL.

FOXP2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FOXP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat




 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-625
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol FOXP2
Antigen name Forkhead Box P2
Reactivity Human, Rat
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms trinucleotide repeat-containing gene 10 protein, CAGH44, SPCH1, trinucleotide repeat containing 10, CAG repeat protein 44, forkhead/winged-helix transcription factor, TNRC10, forkhead box protein P2
Concentration 500 ug/mL
Immunogen FOXP2 antibody was raised against a 14 amino acid synthetic peptide near the internal region of FOXP2.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo
 

Nový vzhľad a obnovený záväzok

Postavené na funkciách a zdrojoch, ktoré poznáte, optimalizované s inováciami od spoločnosti Avantor®. Naša nová značka symbolizuje agilitu a našu schopnosť prispôsobiť sa potrebám zákazníkov.

ĎALŠIE INFORMÁCIE ZAČAŤ NAKUPOVAŤ