Tlačiť

Anti-FOXQ1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-628EA 614.75 EUR
PRSI45-628
Anti-FOXQ1 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 50 kDa band (faint) observed in H460 lysate (predicted MW of 46 kDa according to NP_150285). Recommended for use at 1-3 μg/mL.

FOXQ1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FOXQ1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXQ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR