Tlačiť

Anti-FOSL1 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
PRSI45-631EA 616.08 EUR
PRSI45-631
Anti-FOSL1 Goat Polyclonal Antibody
Protilátky
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 26-28 kDa band observed in human bone marrow, breast and placenta lysates (calculated MW of 29.4 kDa according to NP_005429.1). Recommended concentration: 0.1-0.3 μg/mL.

FRA1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FRA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR