Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-FZD7 Goat Polyclonal Antibody

Dodávateľ: ProSci Inc.
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 60 kDa band observed in lysates of cell lines HEK293 and HepG2 (calculated MW of 63.6 kDa according to NP_003498.1). Recommended concentration: 0.3-1 μg/mL.

Frizzled 7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Frizzled 7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Antigen: FZD7
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Goat
Reactivity: Human

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení


Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG PRSI45-635
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol FZD7
Antigen name Frizzled 7
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
Antigen synonyms fz-7, Fz-7, FzE3, hFz7
Concentration 500 ug/mL
Immunogen Frizzled 7 antibody was raised against a 14 amino acid synthetic peptide near the C-Terminus of Frizzled 7.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo