Microplate luminometers, Luminoskan™
Dodávateľ: Thermo Fisher Scientific
LABL5300332EA
29366.15
EUR
LABL5300332
LABL5300331
Microplate luminometers, Luminoskan™
Useful for a variety of luminometric research applications, the Thermo Scientific™ Luminoskan™ Microplate Luminometer is a compact and robust instrument offering plate format versatility, fast reading speeds, and up to three dispensers. Luminoskan™ offers simplicity and reliability for increased efficiency of workflows.
- High sensitivity for both top and bottom reading
- Fast reading speed essential for kinetic applications
- Onboard dispensers for precise initiation of kinetic reactions
- Powerful and easy to use Thermo Scientific™ SkanIt™ software
The use of fibreless direct illumination optics for both top and bottom reading enables high sensitivity, a wide dynamic range, and low cross-talk for accurate and precise results in luminometric assays. The Luminoskan luminometer is certified as DLReady for excellent sensitivity.
The Luminoskan™ Microplate Luminometer is capable of reading a 96-well plate in just 18 seconds, essential for kinetic applications such as enzyme kinetic studies and phagocytosis assays.
The Luminoska™n Microplate Luminometer can be equipped with up to three reagent dispensers* for use during fast kinetic assays, such as Ca2+ flux assays. The onboard dispensers support simultaneous dispensing and reading, enabling the monitoring of fast kinetic measurements form the very start of the reaction. Very low dead volume and backflush capability support conservation of reagents.
With a visual workflow setup, SkanIt™ software is quick and easy to learn. It offers easy data analysis (with a comprehensive selection of built-in calculations, including blank subtraction, standard curve creation, classification, quality control, kinetic analysis) and exporting capabilities. Each user can install SkanIt™ software on his/her own computer to set up assays and analyse data.
The Luminoskan™ Microplate Luminometer is capable of reading a 96-well plate in just 18 seconds, essential for kinetic applications such as enzyme kinetic studies and phagocytosis assays.
The Luminoska™n Microplate Luminometer can be equipped with up to three reagent dispensers* for use during fast kinetic assays, such as Ca2+ flux assays. The onboard dispensers support simultaneous dispensing and reading, enabling the monitoring of fast kinetic measurements form the very start of the reaction. Very low dead volume and backflush capability support conservation of reagents.
With a visual workflow setup, SkanIt™ software is quick and easy to learn. It offers easy data analysis (with a comprehensive selection of built-in calculations, including blank subtraction, standard curve creation, classification, quality control, kinetic analysis) and exporting capabilities. Each user can install SkanIt™ software on his/her own computer to set up assays and analyse data.
Applications include: Reporter gene assays, immunoassays with luminescent substrates, cell proliferation and cytotoxicity assays, intracellular Ca2+ assays, ATP assays, phagocytosis assays, reactive oxygen assays, microbiological assays, enzyme assays, BRET assays, ADMEtox.
Certifikácie: This instrument has passed validation at Promega Corporation to be certified as DLReady. In addition, the Luminoskan Microplate Luminometer is validated for Thermo Scientific Pierce luciferase assays.
Objednávacie informácie: Includes SkanIt™ PC Software. Filters available upon request.
Pozor: *Upgrade to third dispenser performed by service technician.
New Product
Limited Time Sale
Select items below are currently on sale
Zadať detaily certifikátu
Zavolať a opýtať sa na ceny
Learn more
About VWR
Item(s) Added to Cart