Tlačiť
Human peripheral blood serum
Dodávateľ: Charles River Laboratories Cell Solutions
PB007C-1EA 0 EUR
HCMSPB007C-1 HCMSPB007C-2 HCMSPB007F-1
Human peripheral blood serum
Serum is the fluid portion of the blood obtained after removal of the fibrin clot and blood cells, distinguished from the plasma in circulating blood. Human peripheral blood serum is collected in FDA-registered collection centres from IRB consented healthy human volunteer donors.
  • Test negative for Hepatitis B, Hepatitis C, HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV Nucleic Acid, WNV Nucleic Acid, Trypanasoma cruzi and Syphilis
  • Available fresh or frozen
Storage: Store frozen product at −80 °C. Fresh product must be used immediately or refrigerated upon receipt.
Pozor: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.