Tlačiť
Human serum, herpes simplex virus 2 (HSV 2)
Dodávateľ: Charles River Laboratories Cell Solutions
PB007C-1-HSV2EA 0 EUR
PB007C-1-HSV2
Human serum, herpes simplex virus 2 (HSV 2)
Séra
Human serum used for researching the causes, diagnosis, supportive care and treatment of herpes simplex virus 2 (HSV 2). Collected from IRB-consented donors diagnosed with HSV 2.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Store at −80 °C.
Pozor: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.