Tlačiť
Human mononuclear cells, psoriasis (PSO)
Dodávateľ: Charles River Laboratories Cell Solutions
PB009C-1-PSOEA 0 EUR
PB009C-1-PSO
Human mononuclear cells, psoriasis (PSO)
Buňky
Psoriasis (PSO) is isolated from mononuclear cells collected from IRB-consented donors diagnosed with psoriasis (PSO).
  • Used for researching the causes, diagnosis, care and treatment of psoriasis (PSO)
  • Cryopreserved in CryoStor® CS10 media
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Either prepare cells for long term storage in liquid nitrogen vapour phase or thaw for use. Storage in liquid phase nitrogen is NOT recommended. Short term storage of cells (less than 2 weeks) at –80 °C is acceptable, but should be minimised to ensure maximum stability. Once thawed, samples must be used immediately.
Pozor: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.