Tlačiť
Human mononuclear cells, Hepatitis B virus (HBV)
Dodávateľ: Charles River Laboratories Cell Solutions
PB009C-1-HBVEA 894.23 EUR
HCMSPB009C-1-HBV
Human mononuclear cells, Hepatitis B virus (HBV)
Mononuclear cells used for researching the causes, diagnosis, supportive care and treatment of the Hepatitis B Virus (HBV). Patient samples can be used by researchers to investigate biomarker and diagnostic development, pre-clinical research validation, and drug toxicity and efficacy. Human mononuclear cells are collected from human volunteer donors diagnosed with Hepatitis B virus (HBV) under IRB approved informed consent by whole blood or leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol. Mononuclear cells are purified using density gradient centrifugation techniques.
  • Cryopreserved in CryoStor® CS10 media
Storage: Either prepare cells for long term storage in liquid nitrogen vapour phase or thaw for use. Storage in liquid phase nitrogen is NOT recommended. Short term storage of cells (less than 2 weeks) at –80 °C is acceptable, but should be minimised to ensure maximum stability. Once thawed, samples must be used immediately.
Pozor: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices. And storage in liquid phase nitrogen is not recommended.