Tlačiť
Ultra-low temperature freezers, Revco™ RDE
Dodávateľ: Thermo Fisher Scientific
Revco®
471-1244EA 26001.24 EUR
471-1244 471-1242 471-1243 471-1241
Ultra-low temperature freezers, Revco™ RDE
Mrazáky Boxy hlubokomrazící
Thermo Scientific™ Revco™ RDE Series ultra-low freezers feature four upright models, maximizing storage capacity from 30,000 up to 60,000 2 ml vials. New sustainability features include natural refrigerants and water-blown foam, compliant with many sustainability standards.
  • Easy-to-use set-up wizard, event log, temperature mapping and data downloads
  • Capacitive touch even when using latex gloves
  • Touch-screen datalogging and temperature mapping eliminates the need for a separate chart recorder
  • Built-in USB port for exporting data
  • Advanced security features: alarm settings, password-protected controller, padlock-compatible doors and optional access card entry
  • Ergonomic design: comfortable door handle, eye-level touch-screen and shelf-level placement that do not require floor-level loading of racks
  • Five inner doors minimize ambient temperature during sample retrieval
  • Low peak variation helps ensure sample protection throughout entire cabinet
  • Fast door opening recovery in as little as 14 minutes
  • 0,93 °C steady-state temperature variation over time
  • Long warm-up time from −80 to −50 °C in 5 hours
Limit environmental impact and lower HVAC costs:
Uses as low as 11,2 kWh/day, depending on size
Less heat emission lowers HVAC costs
Natural refrigerants compliant with EU F-Gas regulation: R170 and R290
Environmentally-friendly, water-blown foam insulation
Manufactured in an award-winning, zero-waste to landfill facility
Four capacities: 300, 400, 500 and 600 2-inch (5 cm) box
Temperature: −50...−86 °C.
Informácie o príslušenstve: Field installed options and accessories must be installed by a qualified professional.