Tlačiť
Conductivity cell for ultra-pure water
Dodávateľ: WTW
663-0066EA 0 EUR
663-0066
Conductivity cell for ultra-pure water
Elektrody Cely vodivostní
Conductivity cell for use with the portable Vario Cond meter.
Filling volume: 17 ml (without sensor)
Immersion depth: 40 mm