Tlačiť
Incubator-clean
Dodávateľ: AppliChem
A5230.5000RF A5230.0500
Incubator-clean
Prostředky čistící Dezinfekční prostředek
Incubator-Clean™ disinfectant solution prevents contamination with, and growth of, fungi (and spores), bacteria (and spores, including Tuberculosis bacteria), viruses (including HIV and Hepatitis B) and mycoplasma in incubators and/or on sterile work benches. The active ingredients are quaternary benzylammonium compounds.
  • Free of mercury, formaldehyde, phenol or alcohol
  • Non corrosive to aluminium, tin-plated iron, chrome, nickel, steel, high grade steel and copper
  • Non toxic
Recommended use: Spray incubators once every 2 weeks; spray sterile benches once a day, or preferably before each laboratory worker begins to use the work area.
Pozor: Use biocides safely. Always read the label and product information before use.
Incubator-Clean™ is a registered trademark of AppliChem GmbH