Tlačiť
Bicinchoninic acid (BCA) protein assay
Dodávateľ: PanReac AppliChem
APLIA7787.0500
Bicinchoninic acid (BCA) protein assay
Bicinchoninic Acid Protein Assay kit is a formulation based on bicinchoninic acid for the rapid and sensitive detection and quantitation of total protein content.
  • Photometric method (reading at 562 nm)
  • More sensitive than Lowry and less subject to interferences
  • Linear working range for BSA is 20 – 2000 µg/ml for the bicinchoninic acid protein assay
  • Less protein-to-protein variation
  • Reagents are stable two years at room temperature
  • Adaptable to microplate format
The Bicinchoninic acid method is faster and easier than Lowry, with much greater tolerance to interference from non-ionic detergents and buffer salts.
Informace o dodávce: Sufficient for determination in either 500 test tubes or 5000 micro plate assays.