Tlačiť
ESD safety shoes, lace-up, Biomex Dynamics, Reaction Grey Low, 38320
Dodávateľ: Elten
ELTN72271/45EA 108.06 EUR
ELTN72271/45 ELTN72271/42
ESD safety shoes, lace-up, Biomex Dynamics, Reaction Grey Low, 38320
Metal-free ESD safety shoes made from waxy cowhide leather, full, padded bellows tongue and an extremely flexible, ergonomically S-shaped dual-density TPU/PU sole BIOMEX DYNAMICS.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO black
  • Anatomically shaped insole and ankle support for improved stability
  • Abrasion-proof scuff cap
  • Metal-free, puncture resistant midsole
  • Composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifikácie: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC