Tlačiť
Bead dispensers for 45 tubes
IBSA45-2-50 IBSA45-2-100 IBSA45-2-250 IBSA45-2-1000 IBSA45-2-999
Bead dispensers for 45 tubes
The 45-tube bead dispenser is used to dispense beads in 45 tubes placed in a BioSpec tube holder. Can dispense a wide range of solid quantities in a highly precise and repeatable fashion.
  • Simple, fast and reliable
  • High productivity improvement 60-150 x
  • Easy cleaning with 70% ethanol or soap
  • Sterilisable and not autoclavable
  • Ergonomic use as it removes tedious repetitive tasks
  • Custom made volume plates are available
Material: ABS+PC, nylon, RVS304, anodised aluminum
* Customised size
Informace o dodávce: Supplied with rack, converter and flight case.