Tlačiť

Anti-PCDA5 Mouse Monoclonal Antibody

BSENM-917-200EA 378.8 EUR
BSENM-917-200
Anti-PCDA5 Mouse Monoclonal Antibody
Protocadherin alpha-5 is 1 of 15 tandemly arranged genes within the PCDHA gene cluster. These 15 genes function as 'variable' exons that are individually spliced to a downstream constant region to form distinct PCDHA transcripts. They encode cadherin-like cell surface proteins that are expressed in neurons and are present at synaptic junctions. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain.

This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PCDA5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR