Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

D(+)-Glukóza monohydrát

Dodávateľ: U.S. Pharmacopeia



Vzorec: C₆H₁₂O₆·H₂O
MW: 198,17 g/mol
Teplota tavenia: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00149450
Číslo CAS: 14431-43-7
EINECS: 200-075-1
https://sk.vwr.com/stibo/web/std.lang.all/21/23/2302123.jpg

 

OBJEDNÁVKA

Rozšíriť všetko / Schovať všetko
Pack type Pk Číslo v katalógu VWR Množstvo
Glass bottle for solids 500 mg USPH1181302
Rozšíriť všetko / Schovať všetko
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo