Tlačiť

Agaróza IEF, Electran for electrophoresis

443174YEA 2350.57 EUR
443174Y 443173H
Agaróza IEF, Electran for electrophoresis
Agaróza

Agarose IEF is designed to be free of electroendosmosis which makes it ideal for isoelectric focusing applications with all pH ranges. It is also recommended for use in isotachophoresis.
The agarose forms highly porous gels, which permit rapid focusing of high molecular weight proteins. The agarose solutions can easily be cast into thin gels for improved resolution, the gels rapidly setting at room temperature.


  • DNase and RNase free

Teplota tavenia: 60…90 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00081294
Číslo CAS: 9012-36-6
EINECS: 232-731-8
https://sk.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg

Order Now

Výsledky špecifikačného testu

Electroendosmosis Below detection
Gel strength (1.5 %; water) Min. 500 GCM
SO4 (Sulphate) Max. 0.20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR