Tlačiť
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Dodávateľ: Lonza
LONZ08-238Z10EA 336.28 EUR
LONZ08-238Z10 733-1624 LONZ08-238Z4
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Pufry Pufry pro elektroforézu Pufry do dělícího gelu
AccuGENE™ TBE Buffer Electrophoresis Buffers are formulated for maximum performance and convenience, and are optimized for use with agarose and precast gels.
  • Prepared with 18 megOhm water
  • Filtered using a 0.2-micron filter
  • Product is guaranteed DNase/RNase-free.
0.89 M Tris-borate, 0.02 M EDTA (disodium salt), pH 8.3.