Tlačiť
ESD safety ankle boots, lace-up, uvex 1, 8554
Dodávateľ: Uvex
UVEX8554.2/38 UVEX8554.2/39 UVEX8554.2/47 UVEX8554.2/48 UVEX8554.1/36 UVEX8554.2/45 UVEX8554.2/46 UVEX8554.1/35 UVEX8554.2/43 UVEX8554.2/44 UVEX8554.2/41 UVEX8554.2/42 UVEX8554.3/51 UVEX8554.3/52 UVEX8554.2/40 UVEX8554.3/50 UVEX8554.3/39 UVEX8554.3/48 UVEX8554.2/36 UVEX8554.2/37 UVEX8554.3/49 UVEX8554.3/46 UVEX8554.2/35 UVEX8554.3/47 UVEX8554.3/44 UVEX8554.3/45 UVEX8554.3/42 UVEX8554.3/43 UVEX8554.3/40 UVEX8554.3/41 UVEX8554.4/52 UVEX8554.4/51 UVEX8554.4/50 UVEX8554.1/49 UVEX8554.1/48 UVEX8554.3/37 UVEX8554.4/49 UVEX8554.3/38 UVEX8554.3/35 UVEX8554.4/48 UVEX8554.3/36 UVEX8554.4/47 UVEX8554.1/52 UVEX8554.4/46 UVEX8554.1/51 UVEX8554.4/45 UVEX8554.4/44 UVEX8554.4/43 UVEX8554.4/42 UVEX8554.4/41 UVEX8554.4/40 UVEX8554.1/50 UVEX8554.1/38 UVEX8554.1/37 UVEX8554.2/49 UVEX8554.1/39 UVEX8554.4/39 UVEX8554.1/45 UVEX8554.1/44 UVEX8554.4/38 UVEX8554.4/37 UVEX8554.1/47 UVEX8554.1/46 UVEX8554.4/36 UVEX8554.1/41 UVEX8554.4/35 UVEX8554.1/40 UVEX8554.2/52 UVEX8554.1/43 UVEX8554.1/42 UVEX8554.2/50 UVEX8554.2/51
ESD safety ankle boots, lace-up, uvex 1, 8554
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock-absorbent at heel and front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • Elastic laces for quick, individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Flexible, metal-free, puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Width: 10, 11, 12, 14
Certifikácie: EN 61340 (ESD); DGUV 112-191; EN ISO 20345: 2011, S3, SRC