Penicillin/Streptomycin/Glutamine 100X
Dodávateľ: BIOWEST
L0014-100EA
29.61
EUR
L0014-100
Penicillin/Streptomycin/Glutamine 100X
Penicillin/Streptomycin/Glutamine
Glutamine penicillin streptamycin acts against gram-negative and gram-positive bacteria.
- Storage temperature of –20 °C
- Shelf life of 24 months
- Endotoxin: <10 EU/ml
Used in cell culture applications at 10 ml/l. This concentration is for tissue culture media containing serum; serum-free media generally require lower concentration.
Pozor: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.