Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Etylstearát ≥99%

Dodávateľ: Alfa Aesar

Purity: 99%
CAS No.: 111-61-5
Molecular Formula: C20H40O2
Formula Weight: 312.54
Flash Point: 110°(230°F)
Melting Point: 33-36°
Boiling Point: 213-215°/15mm
Beilstein Reference: 1788183
MDL No.: MFCD00009006


Vzorec: C₂₀H₄₀O₂
MW: 312,54 g/mol
Teplota varu: 365 °C (1013 hPa)
Teplota tavenia: 31 °C
Hustota: 0,8973 g/cm³ (25 °C)
Storage Temperature: Refrigerator
Číslo CAS: 111-61-5
EINECS: 203-887-4


 

OBJEDNÁVKA

Rozšíriť všetko / Schovať všetko
Pack type Pk Číslo v katalógu VWR Množstvo
Glass bottle for solids 5 g A16807.06
Rozšíriť všetko / Schovať všetko
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Melting point 33-36°
Formula weight 312.54
CAS No. 111-61-5
Purity 99%
Flash point >110°(230°F)
Molecular formula C20H40O2
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo