Chcete sa opäť prihlásiť?
Metylvalerát
Dodávateľ: Acros Organics
Vzorec: C₆H₁₂O₂
MW: 116,16 g/mol
Teplota varu: 126,5 °C (1013 hPa)
Teplota tavenia: -91 °C
Hustota: 0,89 g/cm³ (20 °C)
Storage Temperature: Ambient
Číslo CAS: 624-24-8
EINECS: 210-838-0
UN: 3272
ADR: 3,II
OBJEDNÁVKA
Rozšíriť všetko / Schovať všetko
Pack type Pk Číslo v katalógu VWR Množstvo
Glass bottle for solids 25 g ACRO415050250
Rozšíriť všetko / Schovať všetko
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo