Tlačiť

Ethyl myristate

Dodávateľ: EHRENSTORFER GMBH

EHERC17396730EA 69.01 EUR
EHERC17396730
Ethyl myristate
Ethyl myristate
Vzorec: C₁₆H₃₂O₂
Mw: 256,43 g/mol
Hustota: 0,86 g/cm³ (20 °C)
Číslo MDL: MFCD00008984
Číslo CAS: 124-06-1
EINECS: 204-675-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR