Tlačiť
Weighing boats, aluminium
Dodávateľ: Merck
SIALW1126-100EAEA 179.81 EUR
SIALW1126-100EA
Weighing boats, aluminium
Lodičky Lodičky navažovací
Made of aluminium.
  • Lightweight
  • Good chemical resistance
  • Good temperature resistance
Ideal for electrobalance weighing and combustion analysis. Boats are rectangular with flat bottom and tweezer-grip tab for easy handling.