Uhličitan vápenatý, light powder USP, FCC, J.T.Baker®
1301-01EA
220.64
EUR
1301-01
Uhličitan vápenatý, light powder USP, FCC, J.T.Baker®
Uhličitan vápenatý
Vzorec:
CaCO₃ Mw: 100,09 g/mol Teplota tavenia: 825 °C Hustota: 2,93 g/cm³ (20 °C) Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00010906 Číslo CAS: 471-34-1 EINECS: 207-439-9 Merck Index: 13,01658 |
Výsledky špecifikačného testu
Meets F.C.C. Requirements | |
Meets U.S.P Requirements | |
GMP Manufactured Product | |
Food GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
USP - Assay (CaCO₃) (dried basis) | 98.0 - 100.5 % |
USP - Identification | Passes Test |
USP - Acid-Insoluble Substances | ≤0.2 % |
USP - Fluoride (F) | ≤0.005 % |
USP - Arsenic (As) | ≤3 ppm |
USP - Barium (Ba) | Passes Test |
USP - Iron (Fe) | ≤0.1 % |
USP - Lead (Pb) | ≤3 mg/kg |
USP - Magnesium and Alkali Salts | ≤1.0 % |
USP - Mercury (Hg) | ≤0.5 ppm |
FCC - Assay (CaCO₃) (dried basis) | 98.0 - 100.5 % |
FCC - Identification | Passes Test |
FCC - Loss on Drying | ≤2.0 % |
FCC - Acid-Insoluble Matter | ≤0.2 % |
FCC - Fluoride (F) | ≤0.005 % |
FCC - Arsenic (As) | ≤3 ppm |
FCC - Lead (Pb) | ≤3 mg/kg |
FCC - Magnesium and Alkali Salts | ≤1.0 % |
Bulk Density (lb/cu ft) | 32 - 36 |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Metallic Residues: No metal catalysts or metal reagents, as defined by | |
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of | |
this material. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...