Tlačiť

Aluminium oxide, α-phase

39814.22
Aluminium oxide, α-phase
Aluminium oxide, α-phase
Vzorec: Al₂O₃
Mw: 101,96 g/mol
Číslo MDL: MFCD00003424
Číslo CAS: 1344-28-1
EINECS: 215-691-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR