Tlačiť
miRNA Mimic, MISSION®
Dodávateľ: Merck
SIALHMI0195-5NMOLEA 0 EUR
SIALHMI0195-5NMOL SIALHMI0193-5NMOL SIALHMI0197-5NMOL SIALHMI0198-5NMOL SIALHMI0196-5NMOL SIALHMI0199-5NMOL SIALHMI0194-5NMOL SIALHMI0192-5NMOL SIALHMI0191-5NMOL SIALHMI0190-5NMOL
miRNA Mimic, MISSION®
Činidla pro nukleové kyseliny RNAi
MISSION miRNA mimics are small, double-stranded RNA molecules designed to mimic endogenous mature miRNA molecules when introduced into cells.
  • Optimised and ready for transfection
  • Functionally tested for knockdown efficiency against natural miRNA targets
  • Reduces possible sense strand off target effects
  • Available as a whole human library and individual miRNA targets
miRNA are known to regulate gene expression in a variety of manners, including translational repression, mRNA cleavage and deadenylation.