Tlačiť
Grain sieves, brass
Dodávateľ: Endecotts
ENDE200BGS2.80EA 0 EUR
ENDE200BGS2.80 ENDE200BGS2.50 ENDE200BGS2.20 ENDE200BGS2.25
Grain sieves, brass
These grain sieves are used for testing grains and cereals. Mesh is made of perforated plate with slotted holes.
  • Precision frame ensures consistent nestability
  • Precise aperture
  • Natural fillet for free flowing of sample
  • Totally sealed and no crevice to lose material
  • Serial number ensures full traceability
Certifikácie: ISO 5223.