Tlačiť
Blow lamp, HyperTorch™
Dodávateľ: Camping Gaz
CAMP3000004126EA 39.11 EUR
CAMP3000004126
Blow lamp, HyperTorch™
Ohřívače Hořáky plynové
Hyperformance HyperTorch ™ blowtorch is ideal for all soldering, welding and brazing jobs. They are essential tools due to their versatile capabilities: aside from soldering, they can also be used to light barbecues, defrost pipes, brazing, burning, heating, etc.
  • Effortless upside down working capabilities with only five seconds pre-heat time
  • Ergonomic handle for extra comfort
  • Dissipation holes
  • Stabilizer
  • Fine flame nozzle for precision work
Hyperformance blowtorches offer up to 35% more power than standard butane gas blowtorches. The large range of adaptable burners available also provide the blowtorches with additional capabilities.
Wattage: 2240
Flame temperature: 1930 °C
Gas consumption: 160 g/h