Tlačiť
Premium grade BS3, Pierce™
Dodávateľ: Thermo Scientific
Pierce
PIERPG82083EA 485.57 EUR
PIERPG82083 PIERPG82084
Premium grade BS3, Pierce™
Systémy pro expresi proteinů Činidla pro expresi bílkovin
Premium Grade BS3 provides more clearly defined quality and product support by including increased analytical testing and product characterisation, greater batch-specific information and quality assurance review, extensive lot sample retention, and change control notification.
  • Homobifunctional, non-cleavable, membrane-impermeable, water-soluble crosslinking agent for amines
  • Sufficient for 174 ml of typical 1× (1 mM) reaction solution or for ~50 uses at 2 mg reagent per use
Used for crosslink cell-surface proteins prior to cell lysis and immunoprecipitation, identify receptor-ligand interactions by chemical crosslinking, 'Fix' protein interactions to allow identification of weak or transient protein interactions, identify near-neighbor protein interactions, crosslink proteins to create bioconjugates via single-step reactions, immobilise proteins onto amine-coated surfaces.