Tlačiť
Hotplates, aluminium top, 2200 series
Dodávateľ: Thermo Scientific
BARNHPA2245MQEA 1656.48 EUR
BARNHPA2245MQ
Hotplates, aluminium top, 2200 series
Aluminum top hotplates features a large, aluminum heating surface and thermostatic temperature controls for outstanding temperature uniformity and stability.
  • Cycle light indicates when power is being supplied to heating element
  • Epoxy-painted surface increases chemical resistance in corrosive environments
  • Stainless-steel case provides safe, reliable construction and optimal strength for heavy loads
The stainless-steel case provides safe, reliable construction and optimal strength for heavy loads. The temperature uniformity at 100 °C is ±10 °C.
Ideal for large-volume heating applications requiring precise temperature stability, including acid base digestion, sample drying, general reagent heating, heating TLC plates, and evaporations.
Certifikácie: UL listed.
Pozor: Recommended for use with glass vessels only.