Tlačiť
Welding spectacles, uvex astrospec 2.0
Dodávateľ: Uvex
uvex astrospec 2.0
UVEX9164.143EA 25.54 EUR
UVEX9164.143
Welding spectacles, uvex astrospec 2.0
Brýle
This iconic welding protection eyewear features new filter technology for UV and IR radiation protection.
  • Perfect colour recognition
  • Minimum damage from welding sparks
  • Fully adjustable side arms enable wearers to achieve a customised fit
  • Multi-level side arm inclination and four different side arm lengths
  • uvex ventilation system with integrated brow guard for added comfort
  • Flat-fold design fits easily into breast pocket
The panoramic polycarbonate lens with integrated side shield gives optimum protection of the whole eye. Soft, adjustable nose piece and soft, non-slip ear pieces (uvex duo component technology) for pressure free fit.
Lens coating of uvex infradur plus with grey welding shade 3 tinting.
Certifikácie: EN 166, EN 169