Tlačiť
ESD safety boots, lace-up, uvex motion light, 6984
Dodávateľ: Uvex
uvex motion light
UVEX6984.8/48EA 90.91 EUR
UVEX6984.8/48 UVEX6984.8/40 UVEX6984.8/43 UVEX6984.8/44 UVEX6984.8/38 UVEX6984.8/42 UVEX6984.8/41 UVEX6984.8/45 UVEX6984.8/47 UVEX6984.8/46 UVEX6984.8/39
ESD safety boots, lace-up, uvex motion light, 6984
Obuv
Wide, sports-fit ESD safety boots made from nubuck leather upper, soft padded collar and tongue, breathable distance mesh and an extremely flexible dual density PUR sole (slip-, abrasion- and cut-resistant, resistant to oil and petrol and withstands brief exposure to heat of approx. +120 °C). Free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • Protective steel toecap (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Colour: Black
Width: 11
Certifikácie: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S2, SRC