Tlačiť
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye
Dodávateľ: Biotium
90023.EA 253.2 EUR
90023.
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE)
6-FAM, SE (full name: 6-Carboxyfluorescein, succinimidyl ester, single isomer) is the amine-reactive form of 6-carboxyfluorescein single isomer.
6-FAM SE is an amine-reactive green fluorescent dye widely used for labeling oligonucleotides or other biomolecules that contain a primary or secondary aliphatic amine. The coupling reaction is usually carried out at pH 8-9.5.
  • λEx/λEm (pH 9) = 495/519 nm
  • Yellow-orange solid soluble in DMF or DMSO
  • Store at -20°C and protect from light
Vzorec: C₂₅H₁₅NO₉
Skladovacia teplota: Freezer
Číslo MDL: MFCD00077323
Číslo CAS: 92557-81-8