FURA-2 pentasodium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Dodávateľ: Biotium
Fura-2, pentasodium salt is a ratiometric fluorescent dye that binds to free intracellular calcium. It is membrane-impermeant and may be loaded into cells via microinjection or scrape loading.
- Kd: 145 nM (22°C in pH 7.2 buffer)
- λEx/λEm (no Ca2+) = 363/512 nm; λEx/λEm (high Ca2+) = 335/505 nm
- ε (363 nm, no Ca2+) = 27,000 M-1cm-1; ε (335 nm, high Ca2+) = 35,000 M-1cm-1
- Cell permeability: Membrane impermeant
- Light yellow solid soluble in DMSO
- Store at 4°C and protect from light, especially in solution
Fura-2 fluorescence can be detected using flow cytometry, fluorescence microscopy, or certain fluoresence microplate readers. Upon calcium binding, the fluorescent excitation maximum of the indicator undergoes a blue shift from 363 nm (Ca2+-free) to 335 nm (Ca2+-saturated), while the fluorescence emission maximum is relatively unchanged at ~510 nm. A major use of Fura-2 is in drug discovery, for measuring intracellular calcium mobilization after activation of G protein-coupled receptors (GPCRs) and calcium channels.
Vzorec:
C₂₉H₂₂N₃Na₅O₁₄ Skladovacia teplota: Refrigerator |
Číslo MDL:
MFCD00083328 Číslo CAS: 96314-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...